This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245505_g1
See other SLC2A4 GE Assays ›
Gene Symbol:
Slc2a4
Gene Name:
solute carrier family 2 (facilitated glucose transporter), member 4
Gene Aliases:
Glut-4, Glut4
Chromosome Location:
Chr.11: 69942286 - 69948221 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245505_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc2a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009204.2 NP_033230.2 8 - 9 1227 61
GenBank mRNA AB008453.1 - 8 - 9 1025 61
AK137607.1 - 8 - 9 1212 61
AK138832.1 - 8 - 9 1202 61
AK160882.1 - 7 - 8 1328 61
AK164310.1 - 8 - 9 1202 61
BC014282.1 - 8 - 9 1197 4207674 61
M23383.1 - 8 - 9 1177 61

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc2a4

Gene Ontology Categories:

Function(s) Process(es)

transport
carbohydrate transport
amylopectin biosynthetic process
glucose transport
proton transport
cellular response to insulin stimulus
hexose transmembrane transport
glucose homeostasis
glucose import
brown fat cell differentiation
transmembrane transport
cellular response to hypoxia
cellular response to osmotic stress
cation transmembrane transport
transporter activity
glucose transmembrane transporter activity
insulin-responsive hydrogen:glucose symporter activity
protein binding
transmembrane transporter activity
substrate-specific transmembrane transporter activity
D-glucose transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100028321.mm.1 NM_009204.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028321.mm.1 NM_009204.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100028321.mm.1 TC1100003078.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0