This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243562_g1
See other CENPT GE Assays ›
Gene Symbol:
Cenpt
Gene Name:
centromere protein T
Gene Aliases:
G630055P03Rik
Chromosome Location:
Chr.8: 105844669 - 105855038 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243562_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cenpt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177150.2 NP_796124.1 11 - 12 1348 80
GenBank mRNA AK089172.1 - 11 - 12 1397 80
AK090340.1 - 11 - 12 1349 80
AK143613.1 - 11 - 12 1311 80
AK164039.1 - 7 - 8 949 80
AK167376.1 - 11 - 12 1349 80
BC022690.1 - 2 - 3 208 80
BC080832.1 - 10 - 11 1339 80
BC100321.1 - 5 - 6 2779 80
BC121824.1 - 11 - 12 1349 40107149 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002838.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cenpt

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
cell cycle
chromosome segregation
mitotic nuclear division
chromosome organization
cell division
kinetochore assembly
molecular_function
DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021848.mm.1 NM_177150.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021848.mm.1 NM_177150.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800021848.mm.1 TC0800002838.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0