This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242812_g1
See other PSMC5 GE Assays ›
Gene Symbol:
Psmc5
Gene Name:
protease (prosome, macropain) 26S subunit, ATPase 5
Gene Aliases:
mSUG1
Chromosome Location:
Chr.11: 106256185 - 106263112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242812_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psmc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008950.1 NP_032976.1 4 - 5 272 102
GenBank mRNA AK010505.1 - 4 - 5 292 102
AK012294.1 - 4 - 5 304 102
AK135451.1 - 4 - 5 282 102
AK145703.1 - 4 - 5 276 102
BC030840.1 - 4 - 5 304 3994523 102
Z54219.1 - 4 - 5 272 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001777.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psmc5

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

protein catabolic process
ER-associated ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
positive regulation of inclusion body assembly
positive regulation of proteasomal protein catabolic process
nucleotide binding
receptor binding
protein binding
ATP binding
transcription factor binding
hydrolase activity
ATPase activity
TBP-class protein binding
thyrotropin-releasing hormone receptor binding
proteasome-activating ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100016807.mm.1 NM_008950.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016807.mm.1 NM_008950.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016807.mm.1 TC1100001777.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0