This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242536_g1
See other POLG2 GE Assays ›
Gene Symbol:
Polg2
Gene Name:
polymerase (DNA directed), gamma 2, accessory subunit
Gene Aliases:
-
Chromosome Location:
Chr.11: 106754821 - 106780010 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242536_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015810.2 NP_056625.2 5 - 6 1124 99
NR_027785.1 5 - 6 977 99
GenBank mRNA AF177202.1 - 5 - 6 1029 99
AK041591.1 - 5 - 6 1096 99
AK042777.1 - 5 - 6 935 99
AK164730.1 - 5 - 6 984 99
AK169977.1 - 5 - 6 1366 99
BC119131.1 - 5 - 6 1053 8733880 99
BC144810.1 - 5 - 6 928 99
BE853186.1 - 3 - 4 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003934.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polg2

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
DNA replication
mitochondrial DNA replication
DNA repair
respiratory electron transport chain
mitochondrial DNA metabolic process
mitochondrion morphogenesis
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
identical protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103446 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103445 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103448 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103444 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103447 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103449 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023191 AK169977.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023191 BC119131.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023191 AF177202.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023191 AK041591.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023191 NM_015810.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104473 BC144810.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104473 BE853186.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104473 AK169977.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104473 BC119131.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P104473 NR_027785.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823974 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842052 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113251 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762065 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647836 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158591 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763952 AK076218.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763952 AK154859.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763952 BC020019.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763952 BC022979.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763952 BC096594.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763952 NM_144843.3 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149370 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179697 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165944 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023191 AK169977.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023191 BC119131.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023191 AF177202.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023191 AK041591.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023191 NM_015810.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104473 BC144810.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104473 BE853186.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104473 AK169977.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104473 BC119131.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P104473 NR_027785.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589591 BC144810.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589591 BE853186.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589591 AK169977.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589591 BC119131.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P589591 NR_027785.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103435 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103439 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103453 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103451 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103441 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103443 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28103444 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07480666 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165944 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179697 - -
Whole Mouse Genome G4122A A_52_P589591 BC144810.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P589591 BE853186.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P589591 AK169977.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P589591 BC119131.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P589591 NR_027785.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104473 BC144810.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104473 BE853186.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104473 AK169977.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104473 BC119131.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104473 NR_027785.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165944 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179697 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09219099 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09219098 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09370425 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09548254 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103446 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103447 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103449 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103444 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103448 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28103445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103489 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103492 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103527 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103499 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28103363 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09062322 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103427 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103435 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103420 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09040218 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103443 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103444 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103429 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103449 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103419 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103436 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103415 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103422 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103439 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103454 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103417 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103412 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103438 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09304728 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103418 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103431 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103433 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103453 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103451 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103434 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103421 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103425 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28103430 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10392245 - -
Affymetrix Mouse Gene 1.0 ST Array 10392248 - -
Affymetrix Mouse Gene 1.0 ST Array 10392250 - -
Affymetrix Mouse Gene 1.0 ST Array 10392244 - -
Affymetrix Mouse Gene 1.0 ST Array 10392242 - -
Affymetrix Mouse Gene 1.0 ST Array 10392249 - -
Affymetrix Mouse Gene 1.0 ST Array 10392247 - -
Affymetrix Mouse Gene 1.0 ST Array 10392243 - -
Affymetrix Mouse Gene 1.0 ST Array 10392246 - -
Affymetrix HT MG-430 PM Array Plate 1450816_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10392241 - -
Mouse Genome 430 2.0 Array 1450816_at - -
Mouse Genome 430 2.0 Array 1450816_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 50776_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 50776_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100036888.mm.1 NM_015810.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036888.mm.1 NM_015810.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036888.mm.1 NR_027785.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036888.mm.1 TC1100003934.mm.1 MTA TC
Mouse Expression Set 430 1450816_at - -
Mouse Expression Set 430 1450816_PM_at - -
Murine Genome U74v2 Set 93699_PM_at - -
Murine Genome U74v2 Set 162277_r_at - -
Murine Genome U74v2 Set 162277_PM_r_at - -
Murine Genome U74v2 Set 93699_at - -
Mouse Genome 430A 2.0 Array 1450816_PM_at - -
Mouse Genome 430A 2.0 Array 1450816_at - -
Affymetrix HT Mouse Genome 430A Array 1450816_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1450816_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 5690047 - -
Illumina mouse-6 v1.0 expression beadchip 5690047 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005690047 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772224 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2772224 - -
mouse-6 v1.1 (Illumina) scl39434.9.1_18-S - -
Illumina Sentrix mouse-6.v1 bead array 5690047 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl39434.9.1_18-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690047 - -
MouseRef-8 v2 Expression BeadChip ILMN_2896483 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772224 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2896483 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39434.9.1_18-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header