This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01241747_g1
See other PCIF1 GE Assays ›
Gene Symbol:
Pcif1
Gene Name:
PDX1 C-terminal inhibiting factor 1
Gene Aliases:
2310022K11Rik, C20orf67, F730014I05Rik
Chromosome Location:
Chr.2: 164879368 - 164891437 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01241747_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcif1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146129.3 NP_666241.1 8 - 9 1130 99
GenBank mRNA AK089358.1 - 8 - 9 1131 99
AK150740.1 - 7 - 8 1021 99
BC031431.1 - 8 - 9 1099 4241283 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002538.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcif1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphatase activity
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200020807.mm.1 NM_146129.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020807.mm.1 NM_146129.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200020807.mm.1 TC0200002538.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0