This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01241632_g1
See other CAMP GE Assays ›
Gene Symbol:
Camp
Gene Name:
cathelicidin antimicrobial peptide
Gene Aliases:
CAP18, Cnlp, Cramp, FALL39, MCLP
Chromosome Location:
Chr.9: 109847377 - 109849456 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01241632_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009921.2 NP_034051.2 2 - 3 320 76
GenBank mRNA AK158005.1 - 2 - 3 312 76
BC120752.1 - 2 - 3 317 40129675 76
BC120754.1 - 2 - 3 320 40129677 76
U43409.1 - 2 - 3 313 76
X94353.1 - 2 - 3 312 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003098.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camp

Panther Classification:

Molecular Function -

antimicrobial response protein

Gene Ontology Categories:

Function(s)

positive regulation of protein phosphorylation
innate immune response in mucosa
defense response
positive regulation of cell proliferation
antibacterial humoral response
defense response to bacterium
negative regulation of growth of symbiont in host
negative regulation of growth of symbiont on or near host surface
positive regulation of angiogenesis
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
killing by host of symbiont cells

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900026163.mm.1 NM_009921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900026163.mm.1 NM_009921.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900026163.mm.1 TC0900003098.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0