Assay Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK146459.1 | - | 9 - 10 | 955 | 79 | |
AK158704.1 | - | 9 - 10 | 957 | 79 | ||
BC008130.1 | - | 10 - 11 | 960 | 3498984 | 79 | |
BC034861.1 | - | 9 - 10 | 964 | 79 | ||
X71978.1 | - | 9 - 10 | 979 | 79 | ||
Z67963.1 | - | 9 - 10 | 979 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057102 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057104 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057106 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057103 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057100 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057105 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963737 | Z67963.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963737 | AK158704.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963737 | BC008130.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963737 | BC034861.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963737 | X71978.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572675 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906673 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212749 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716835 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130391 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780784 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106329 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135908 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163620 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963737 | Z67963.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963737 | AK158704.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963737 | BC008130.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963737 | BC034861.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963737 | X71978.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057135 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057090 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26057121 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163620 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135908 | - | - |
Whole Mouse Genome G4122A | A_52_P174313 | Z67963.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P174313 | AK158704.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P174313 | BC008130.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P174313 | BC034861.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P174313 | X71978.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P417477 | Z67963.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P417477 | BC008130.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P417477 | X71978.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P417477 | NM_010241.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P417477 | AK146459.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127897 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163620 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135908 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07349173 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07317282 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07317281 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07317287 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07317280 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07317283 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057100 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057103 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057102 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057104 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057106 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26057105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26057119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07317281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057090 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07349173 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06885686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06701834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06885690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06989090 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07317282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07187340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06806780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06885692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07349169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06885698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07438338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07036051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07324755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26057051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06885689 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10580543 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580544 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580548 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580541 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580542 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580547 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580545 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580538 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580546 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580549 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437767_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430219_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423364_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10580537 | - | - |
Mouse Genome 430 2.0 Array | 1430219_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430219_at | - | - |
Mouse Genome 430 2.0 Array | 1437767_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1423364_a_at | - | - |
Mouse Genome 430 2.0 Array | 1437767_s_at | - | - |
Mouse Genome 430 2.0 Array | 1423364_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14339_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14339_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800020483.mm.1 | NM_010241.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800020483.mm.1 | TC0800002680.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800020482.mm.1 | NM_010241.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800020482.mm.1 | TC0800002680.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800020481.mm.1 | NM_010241.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800020481.mm.1 | TC0800002680.mm.1 | MTA TC |
Mouse Expression Set 430 | 1423364_PM_a_at | - | - |
Mouse Expression Set 430 | 1437767_s_at | - | - |
Mouse Expression Set 430 | 1437767_PM_s_at | - | - |
Mouse Expression Set 430 | 1423364_a_at | - | - |
Murine Genome U74v2 Set | 101892_PM_f_at | - | - |
Murine Genome U74v2 Set | 101894_s_at | - | - |
Murine Genome U74v2 Set | 101894_PM_s_at | - | - |
Murine Genome U74v2 Set | 101893_r_at | - | - |
Murine Genome U74v2 Set | 101893_PM_r_at | - | - |
Murine Genome U74v2 Set | 101892_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1437767_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1423364_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1423364_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1437767_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430219_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1437767_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423364_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1437767_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430219_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423364_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105860692 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6350575 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 105860692 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102480647 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006350575 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2596713 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248486 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1219649 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2596713 | - | - |
mouse-6 v1.1 (Illumina) | scl34502.14.9_18-S | - | - |
mouse-6 v1.1 (Illumina) | scl000558.1_11-S | - | - |
mouse-6 v1.1 (Illumina) | scl0014339.2_163-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102480647 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6350575 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105860692 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0014339.2_163-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102480647 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350575 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105860692 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2596713 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219649 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2596713 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0014339.2_163-S | - | - |
