This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01241197_g1
See other GIT1 GE Assays ›
Gene Symbol:
Git1
Gene Name:
G protein-coupled receptor kinase-interactor 1
Gene Aliases:
Cat-1, p95Cat
Chromosome Location:
Chr.11: 77493412 - 77507775 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01241197_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Git1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004144.1 NP_001004144.1 4 - 5 579 73
GenBank mRNA BC079870.1 - 4 - 5 579 6831910 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001125.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Git1

Gene Ontology Categories:

Function(s) Process(es)

immunological synapse formation
regulation of G-protein coupled receptor protein signaling pathway
regulation of cytokinesis
positive regulation of GTPase activity
ephrin receptor signaling pathway
dendritic spine development
GTPase activator activity
protein binding
small GTPase binding
protein complex binding
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100010204.mm.1 NM_001004144.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100010204.mm.1 NM_001004144.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100010204.mm.1 TC1100001125.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0