This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240678_m1
See other NFATC2 GE Assays ›
Gene Symbol:
Nfatc2
Gene Name:
nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2
Gene Aliases:
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp
Chromosome Location:
Chr.2: 168476410 - 168601657 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240678_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nfatc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037177.2 NP_001032254.1 5 - 6 1889 79
NM_001037178.2 NP_001032255.1 5 - 6 1254 79
NM_001136073.2 NP_001129545.1 5 - 6 1889 79
NM_001291168.1 NP_001278097.1 5 - 6 1774 79
NM_001291169.1 NP_001278098.1 5 - 6 1774 79
NM_001291170.1 NP_001278099.1 5 - 6 1254 79
NM_001291171.1 NP_001278100.1 4 - 5 738 79
NM_001291172.1 NP_001278101.1 5 - 6 1774 79
NM_001291173.1 NP_001278102.1 4 - 5 738 79
NM_001291174.1 NP_001278103.1 5 - 6 1254 79
NM_001291175.1 NP_001278104.1 5 - 6 1369 79
NM_001291176.1 NP_001278105.1 4 - 5 853 79
NM_001291177.1 NP_001278106.1 5 - 6 1369 79
NM_001291178.1 NP_001278107.1 4 - 5 853 79
NM_001291179.1 NP_001278108.1 5 - 6 1369 79
NM_010899.3 NP_035029.2 5 - 6 1889 79
NR_111897.1 4 - 5 738 79
NR_111898.1 4 - 5 853 79
GenBank mRNA AF289078.1 - 5 - 6 1715 79
AK044489.1 - 5 - 6 1775 79
AK083119.1 - 5 - 6 1230 79
AK161174.1 - 3 - 4 464 79
EU887581.1 - 5 - 6 1655 79
EU887582.1 - 5 - 6 1715 79
EU887583.1 - 5 - 6 1252 79
EU887584.1 - 5 - 6 1352 79
EU887585.1 - 4 - 5 736 79
EU887586.1 - 4 - 5 836 79
EU887587.1 - 5 - 6 1655 79
EU887588.1 - 5 - 6 1715 79
EU887589.1 - 5 - 6 1252 79
EU887590.1 - 5 - 6 1352 79
EU887591.1 - 4 - 5 736 79
EU887592.1 - 4 - 5 836 79
EU887599.1 - 5 - 6 1655 79
EU887600.1 - 5 - 6 1715 79
EU887601.1 - 5 - 6 1252 79
EU887602.1 - 5 - 6 1352 79
EU887603.1 - 4 - 5 736 79
EU887604.1 - 4 - 5 836 79
U36575.1 - 5 - 6 1785 79
U36576.1 - 5 - 6 1785 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005218.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nfatc2

Panther Classification:

Molecular Function -

Rel homology transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
positive regulation of gene expression
myotube cell development
cell migration
positive regulation of B cell proliferation
calcineurin-NFAT signaling cascade
response to drug
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
B cell receptor signaling pathway
positive regulation of myoblast fusion
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200044650.mm.1 NM_001037177.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044650.mm.1 NM_001037178.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044650.mm.1 TC0200005218.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200044646.mm.1 NM_001136073.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044646.mm.1 NM_010899.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200044646.mm.1 TC0200005218.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200044649.mm.1 NM_001037177.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044649.mm.1 NM_001037178.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044649.mm.1 TC0200005218.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200044647.mm.1 NM_001136073.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044647.mm.1 NM_010899.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200044647.mm.1 TC0200005218.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0