This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240550_m1
See other NR1H4 GE Assays ›
Gene Symbol:
Nr1h4
Gene Name:
nuclear receptor subfamily 1, group H, member 4
Gene Aliases:
AI957360, Fxr, HRR1, RIP14, Rxrip14
Chromosome Location:
Chr.10: 89454234 - 89533645 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240550_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr1h4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163504.1 NP_001156976.1 5 - 6 1009 110
NM_001163700.1 NP_001157172.1 4 - 5 805 110
NM_009108.2 NP_033134.2 4 - 5 793 110
GenBank mRNA AK132255.1 - 4 - 5 785 110
BC015261.1 - 4 - 5 764 4241705 110
U09416.1 - 4 - 5 762 110
U09417.1 - 5 - 6 973 110
U09418.1 - 5 - 6 970 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002657.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nr1h4

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
nitrogen catabolite activation of transcription from RNA polymerase II promoter
regulation of carbohydrate metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
Notch signaling pathway
bile acid metabolic process
intracellular receptor signaling pathway
regulation of urea metabolic process
intracellular bile acid receptor signaling pathway
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
negative regulation of bile acid biosynthetic process
cellular response to organonitrogen compound
cellular response to bile acid
positive regulation of glutamate metabolic process
positive regulation of ammonia assimilation cycle
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
thyroid hormone receptor activity
zinc ion binding
bile acid binding
bile acid receptor activity
peptide binding
sequence-specific DNA binding
metal ion binding
retinoid X receptor binding
chenodeoxycholic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0