This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240454_g1
See other TLCD2 GE Assays ›
Gene Symbol:
Tlcd2
Gene Name:
TLC domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 75468024 - 75470899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240454_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tlcd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291156.1 NP_001278085.1 3 - 4 490 76
NM_027249.3 NP_081525.1 3 - 4 490 76
GenBank mRNA AK008522.1 - 3 - 4 466 76
BC021945.1 - 3 - 4 461 5098319 76
CV562896.1 - 3 - 4 491 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004270.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tlcd2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926227 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926233 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926226 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926231 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926232 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926234 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926230 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P116326 AK008522.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328622 BC021945.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328622 NM_027249.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328622 BC021945.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328622 NM_027249.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732497 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125614 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328622 BC021945.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328622 NM_027249.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328622 BC021945.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328622 NM_027249.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P116326 AK008522.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926273 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926260 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926257 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926275 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926276 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926277 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926249 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926284 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926266 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926279 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926250 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926285 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926274 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926290 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926264 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926253 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926262 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926282 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926258 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926280 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926270 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926267 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926255 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926292 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926247 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926288 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125614 - -
Whole Mouse Genome G4122A A_51_P328622 BC021945.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P328622 NM_027249.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P328622 BC021945.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P328622 NM_027249.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116326 AK008522.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125614 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09582297 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09222821 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09582270 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09222826 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09222818 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09582289 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09582275 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09582284 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09222820 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926231 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926227 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926230 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926234 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926233 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926232 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926226 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926257 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926232 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926306 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926204 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926226 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926242 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926200 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926307 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926216 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926219 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926202 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926255 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926211 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926245 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926282 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926245 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926302 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926282 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926247 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926274 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09582280 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09030276 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926279 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926217 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926234 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09401456 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926240 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926275 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926294 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09222806 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09477157 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926270 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926306 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926261 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09582287 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926205 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926296 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926258 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09739861 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926224 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926307 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926223 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09401455 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09026205 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926237 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926216 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926257 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926308 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926211 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926255 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09693308 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926298 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926210 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926238 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926213 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926300 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926266 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926250 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09582284 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09582264 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09401460 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09222808 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926232 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926277 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926287 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926264 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926293 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926284 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926260 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926267 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926262 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926242 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27926249 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10378575 - -
Affymetrix Mouse Gene 1.0 ST Array 10378578 - -
Affymetrix Mouse Gene 1.0 ST Array 10378576 - -
Affymetrix Mouse Gene 1.0 ST Array 10378574 - -
Affymetrix Mouse Gene 1.0 ST Array 10378577 - -
Affymetrix Mouse Gene 1.0 ST Array 10378573 - -
Affymetrix HT MG-430 PM Array Plate 1432282_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10378572 - -
Mouse Genome 430 2.0 Array 1432282_PM_a_at - -
Mouse Genome 430 2.0 Array 1432282_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 380712_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 380712_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100009637.mm.1 NM_027249.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100009637.mm.1 TC1100004270.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100009633.mm.1 NM_027249.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100009633.mm.1 TC1100004270.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100040865.mm.1 NM_027249.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040865.mm.1 TC1100004270.mm.1 MTA TC
Mouse Expression Set 430 1432282_a_at - -
Mouse Expression Set 430 1432282_PM_a_at - -
Mouse Genome 430A 2.0 Array 1432282_a_at - -
Mouse Genome 430A 2.0 Array 1432282_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1432282_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1432282_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 454605 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100520369 - -
MouseWG-6 v2 Expression BeadChip 101500609 - -
Illumina mouse-6 v1.0 expression beadchip 101500609 - -
Illumina mouse-6 v1.0 expression beadchip 100520369 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1249828 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2671810 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1213090 - -
mouse-6 v1.1 (Illumina) scl0380712.4_31-S - -
Illumina Sentrix mouse-6.v1 bead array 101500609 - -
Illumina Sentrix mouse-6.v1 bead array 100520369 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 101500609 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 100520369 - -
MouseRef-8 v2 Expression BeadChip ILMN_2814385 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213090 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2814385 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0