This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240379_g1
See other CRHR1 GE Assays ›
Gene Symbol:
Crhr1
Gene Name:
corticotropin releasing hormone receptor 1
Gene Aliases:
CRFR1, Crhr
Chromosome Location:
Chr.11: 104131998 - 104175518 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240379_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crhr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007762.4 NP_031788.1 9 - 10 1032 65
GenBank mRNA AB375515.1 - 9 - 10 842 65
AF369656.1 - 1 - 2 76 65
AF483484.1 - 9 - 10 842 65
AF483485.1 - 9 - 10 842 65
AK144430.1 - 9 - 10 1100 65
AK158806.1 - 9 - 10 1033 65
BC103675.1 - 9 - 10 860 40046992 65
BC104735.1 - 10 - 11 860 40046995 65
BC105673.1 - 10 - 11 936 40046994 65
BC105675.1 - 9 - 10 860 40046993 65
FJ668671.1 - 8 - 9 722 65
X72305.1 - 9 - 10 873 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001723.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crhr1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
memory
feeding behavior
visual learning
hormone-mediated signaling pathway
regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
adrenal gland development
positive regulation of cAMP biosynthetic process
epithelial cell differentiation
negative regulation of epinephrine secretion
locomotory exploration behavior
fear response
positive regulation of mast cell degranulation
behavioral response to cocaine
behavioral response to ethanol
regulation of synaptic plasticity
behavioral response to pain
corticotropin secretion
response to electrical stimulus
general adaptation syndrome, behavioral process
long-term synaptic potentiation
cellular response to corticotropin-releasing hormone stimulus
negative regulation of neuron death
negative regulation of voltage-gated calcium channel activity
negative regulation of feeding behavior
regulation of corticosterone secretion
G-protein alpha-subunit binding
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
corticotrophin-releasing factor receptor activity
protein complex binding
peptide binding
corticotropin-releasing hormone receptor activity
corticotropin-releasing hormone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100016274.mm.1 NM_007762.5 Refseq
Affymetrix Mouse Clariom S Array PSR1100016274.mm.1 TC1100001723.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0