This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240273_m1
See other CLGN GE Assays ›
Gene Symbol:
Clgn
Gene Name:
calmegin
Gene Aliases:
4930459O04Rik, AI528775, Cln
Chromosome Location:
Chr.8: 83389826 - 83428552 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240273_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clgn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009904.2 NP_034034.2 6 - 7 718 109
GenBank mRNA AK019534.1 - 6 - 7 632 109
AK034525.1 - 6 - 7 619 109
BC050767.1 - 6 - 7 718 6771060 109
D14117.1 - 6 - 7 731 109
D86323.1 - 6 - 7 731 109
U08373.1 - 6 - 7 731 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000923.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clgn

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
protein complex assembly
multicellular organism development
spermatogenesis
single fertilization
binding of sperm to zona pellucida
cell differentiation
meiotic cell cycle
calcium ion binding
protein binding
protein binding involved in protein folding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006822.mm.1 NM_009904.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800006822.mm.1 TC0800000923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006820.mm.1 NM_009904.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800006820.mm.1 TC0800000923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006821.mm.1 NM_009904.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800006821.mm.1 TC0800000923.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0