This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240266_g1
See other ADGRE5 GE Assays ›
Gene Symbol:
Adgre5
Gene Name:
adhesion G protein-coupled receptor E5
Gene Aliases:
AA409984, Cd97, TM7LN1
Chromosome Location:
Chr.8: 83723250 - 83741311 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240266_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgre5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163030.1 NP_001156502.1 5 - 6 683 79
NM_011925.2 NP_036055.2 6 - 7 818 79
GenBank mRNA CA319344.1 - 4 - 5 559 79
Y18365.1 - 6 - 7 676 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002558.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgre5

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of synapse assembly
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_001163029.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_001163029.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_001163030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_001163030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_001163031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_001163031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_011925.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 NM_011925.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019492.mm.1 TC0800002558.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0