This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239974_m1
See other L3MBTL1 GE Assays ›
Gene Symbol:
L3mbtl1
Gene Name:
l(3)mbt-like (Drosophila)
Gene Aliases:
C630004G01, L3mbtl, mKIAA0681
Chromosome Location:
Chr.2: 162942350 - 162977806 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239974_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: L3mbtl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081338.1 NP_001074807.1 8 - 9 1061 64
GenBank mRNA AK049859.1 - 8 - 9 1055 64
AK220350.1 - 6 - 7 809 64
BC116639.1 - 8 - 9 1125 40061282 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002475.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of mitotic nuclear division
chromatin modification
regulation of gene expression, epigenetic
regulation of megakaryocyte differentiation
negative regulation of transcription, DNA-templated
chromatin binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
nucleosome binding
nucleosomal histone binding
SAM domain binding
methylated histone binding
histone binding
identical protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200020148.mm.1 NM_001081338.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020148.mm.1 NM_001081338.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020148.mm.1 TC0200002475.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0