This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239776_m1
See other COPB2 GE Assays ›
Gene Symbol:
Copb2
Gene Name:
coatomer protein complex, subunit beta 2 (beta prime)
Gene Aliases:
AI256832
Chromosome Location:
Chr.9: 98563731 - 98588375 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239776_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Copb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015827.2 NP_056642.1 4 - 5 446 105
GenBank mRNA AK088064.1 - 4 - 5 447 105
AK153352.1 - 4 - 5 447 105
AK188271.1 - 2 - 3 203 105
AK192657.1 - 2 - 3 202 105
BC006675.1 - 4 - 5 433 3499585 105
BC010842.1 - 4 - 5 436 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001236.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Copb2

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
intra-Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
toxin transport
protein kinase C binding
structural molecule activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900009681.mm.1 NM_015827.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009681.mm.1 NM_015827.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900009681.mm.1 TC0900001236.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900009680.mm.1 NM_015827.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900009680.mm.1 NM_015827.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900009680.mm.1 TC0900001236.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0