This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239243_g1
See other SSH3 GE Assays ›
Gene Symbol:
Ssh3
Gene Name:
slingshot homolog 3 (Drosophila)
Gene Aliases:
BC028922, SSH-3, mSSH-3L
Chromosome Location:
Chr.19: 4261668 - 4270276 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239243_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssh3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198113.2 NP_932781.1 7 - 8 876 64
GenBank mRNA AB099289.1 - 7 - 8 874 64
AK038981.1 - 6 - 7 1448 64
AK148293.1 - 7 - 8 869 64
AK164662.1 - 6 - 7 574 64
AK166342.1 - 7 - 8 882 64
BC028922.1 - 7 - 8 876 3987714 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000913.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssh3

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
regulation of actin polymerization or depolymerization
regulation of lamellipodium assembly
dephosphorylation
regulation of axonogenesis
DNA binding
actin binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007946.mm.1 NM_198113.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007946.mm.1 NM_198113.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007946.mm.1 TC1900000913.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0