This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239142_g1
See other DPP3 GE Assays ›
Gene Symbol:
Dpp3
Gene Name:
dipeptidylpeptidase 3
Gene Aliases:
4930533O14Rik, C86324
Chromosome Location:
Chr.19: 4907229 - 4928287 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239142_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133803.2 NP_598564.2 8 - 9 1005 96
GenBank mRNA AK031020.1 - 8 - 9 970 96
AK039975.1 - 8 - 9 966 96
AK087994.1 - 8 - 9 969 96
AK149853.1 - 8 - 9 970 96
AK150101.1 - 8 - 9 965 96
AK150126.1 - 8 - 9 959 96
AK166842.1 - 8 - 9 970 96
AK168195.1 - 8 - 9 958 96
AK207387.1 - 1 - 2 41 96
AK208198.1 - 1 - 2 41 96
BC004600.1 - 7 - 8 947 3584924 96
BC066817.1 - 8 - 9 956 30429618 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000941.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dpp3

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
aminopeptidase activity
peptidase activity
metallopeptidase activity
dipeptidyl-peptidase activity
zinc ion binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900008317.mm.1 NM_133803.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008317.mm.1 NM_133803.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900008317.mm.1 TC1900000941.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0