Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024657.mm.1 |
NM_172461.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024657.mm.1 |
NM_172461.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024657.mm.1 |
TC0900002990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024659.mm.1 |
NM_172461.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024659.mm.1 |
NM_172461.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024659.mm.1 |
TC0900002990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024660.mm.1 |
NM_172461.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024660.mm.1 |
NM_172461.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024660.mm.1 |
TC0900002990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024656.mm.1 |
NM_172461.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024656.mm.1 |
NM_172461.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024656.mm.1 |
TC0900002990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024661.mm.1 |
NM_172461.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024661.mm.1 |
NM_172461.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024661.mm.1 |
TC0900002990.mm.1 |
MTA TC |