This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01237124_m1
See other IL1RL2 GE Assays ›
Gene Symbol:
Il1rl2
Gene Name:
interleukin 1 receptor-like 2
Gene Aliases:
AI481289, AW551444, IL-1Rrp2
Chromosome Location:
Chr.1: 40324441 - 40367585 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01237124_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il1rl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133193.3 NP_573456.1 2 - 3 261 157
GenBank mRNA AF284433.1 - 2 - 3 266 157
BC117797.2 - 2 - 3 262 40092208 157

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF284433.1, BC117797.2, NM_133193.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il1rl2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

immune system process
inflammatory response
signal transduction
cytokine-mediated signaling pathway
positive regulation of interleukin-6 production
innate immune response
positive regulation of T cell differentiation
regulation of inflammatory response
cytokine receptor activity
interleukin-1 receptor activity
interleukin-1, Type I, activating receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0