This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01237033_g1
See other GDPD2 GE Assays ›
Gene Symbol:
Gdpd2
Gene Name:
glycerophosphodiester phosphodiesterase domain containing 2
Gene Aliases:
9130017L10Rik, Gde3, Obdpf
Chromosome Location:
Chr.X: 100729836 - 100738900 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01237033_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gdpd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB048364.1 - 2 - 3 313 76
AK018634.1 - 2 - 3 140 76
AK031379.1 - 2 - 3 376 76
AK086508.1 - 2 - 3 306 76
BC038274.1 - 2 - 3 383 4457335 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000961.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

phosphodiesterase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006245.mm.1 NM_023608.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006245.mm.1 TC0X00000961.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006246.mm.1 NM_023608.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006246.mm.1 TC0X00000961.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0