This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236986_m1
See other PDK3 GE Assays ›
Gene Symbol:
Pdk3
Gene Name:
pyruvate dehydrogenase kinase, isoenzyme 3
Gene Aliases:
2610001M10Rik, AI035637
Chromosome Location:
Chr.X: 93764616 - 93832095 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236986_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdk3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145630.2 NP_663605.1 5 - 6 730 70
GenBank mRNA AK075879.1 - 5 - 6 726 70
AK075985.1 - 5 - 6 786 70
AK088478.1 - 5 - 6 811 70
AK132024.1 - 5 - 6 754 70
AK169874.1 - 5 - 6 811 70
BC008126.1 - 5 - 6 730 3500763 70
CT010338.1 - 5 - 6 587 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002637.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdk3

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glucose metabolic process
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
phosphorylation
peptidyl-serine phosphorylation
peroxisome proliferator activated receptor signaling pathway
cellular response to glucose stimulus
cellular response to fatty acid
hypoxia-inducible factor-1alpha signaling pathway
regulation of reactive oxygen species metabolic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
ATP binding
kinase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00016486.mm.1 NM_145630.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016486.mm.1 NM_145630.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016486.mm.1 TC0X00002637.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0