This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236503_m1
See other IGHMBP2 GE Assays ›
Gene Symbol:
Ighmbp2
Gene Name:
immunoglobulin mu binding protein 2
Gene Aliases:
AEP, Catf1, RIPE3b1, Smbp-2, Smbp2, Smubp2, nmd, sma
Chromosome Location:
Chr.19: 3259076 - 3283010 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236503_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ighmbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009212.2 NP_033238.2 7 - 8 1287 61
GenBank mRNA AK142456.1 - 7 - 8 1091 61
AK153763.1 - 7 - 8 1248 61
AK154805.1 - 7 - 8 1284 61
AK155899.1 - 7 - 8 1284 61
BC052025.1 - 6 - 7 1159 61
BC060085.1 - 6 - 7 1159 61
BC066215.1 - 7 - 8 1230 61
BC083142.1 - 7 - 8 1092 61
L10075.1 - 7 - 8 1183 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000878.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ighmbp2

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
translation
spinal cord motor neuron differentiation
DNA duplex unwinding
protein homooligomerization
tRNA binding
nucleotide binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
RNA binding
helicase activity
ATP binding
DNA-dependent ATPase activity
transcription factor binding
RNA-dependent ATPase activity
zinc ion binding
hydrolase activity
ATP-dependent 5'-3' RNA helicase activity
ribosome binding
ATP-dependent 5'-3' DNA helicase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007538.mm.1 NM_009212.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007538.mm.1 NM_009212.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007538.mm.1 TC1900000878.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900007540.mm.1 NM_009212.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007540.mm.1 NM_009212.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007540.mm.1 TC1900000878.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0