This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236501_g1
See other IGHMBP2 GE Assays ›
Gene Symbol:
Ighmbp2
Gene Name:
immunoglobulin mu binding protein 2
Gene Aliases:
AEP, Catf1, RIPE3b1, Smbp-2, Smbp2, Smubp2, nmd, sma
Chromosome Location:
Chr.19: 3259076 - 3283010 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236501_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ighmbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009212.2 NP_033238.2 3 - 4 675 68
GenBank mRNA AK142456.1 - 3 - 4 479 68
AK153763.1 - 3 - 4 636 68
AK154805.1 - 3 - 4 672 68
AK155899.1 - 3 - 4 672 68
BC066215.1 - 3 - 4 618 68
BC083142.1 - 3 - 4 480 68
BC133681.1 - 3 - 4 483 68
L10075.1 - 3 - 4 571 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ighmbp2

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
translation
spinal cord motor neuron differentiation
DNA duplex unwinding
protein homooligomerization
tRNA binding
nucleotide binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
RNA binding
helicase activity
ATP binding
DNA-dependent ATPase activity
transcription factor binding
RNA-dependent ATPase activity
zinc ion binding
hydrolase activity
ATP-dependent 5'-3' RNA helicase activity
ribosome binding
ATP-dependent 5'-3' DNA helicase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0