This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236406_m1
See other NFYB GE Assays ›
Gene Symbol:
Nfyb
Gene Name:
nuclear transcription factor-Y beta
Gene Aliases:
AA985999, Cbf-A
Chromosome Location:
Chr.10: 82748700 - 82764148 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236406_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nfyb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010914.2 NP_035044.1 5 - 6 644 88
GenBank mRNA AK010762.1 - 5 - 6 624 88
AK011792.1 - 5 - 6 563 88
AK079209.1 - 5 - 6 599 88
AK146168.1 - 5 - 6 646 88
BC010719.1 - 5 - 6 585 2811674 88
X55316.1 - 5 - 6 640 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002572.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nfyb

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein complex binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
repressing transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000019385.mm.1 NM_010914.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000019385.mm.1 NM_010914.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000019385.mm.1 TC1000002572.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0