This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01236114_m1
See other EPAS1 GE Assays ›
Gene Symbol:
Epas1
Gene Name:
endothelial PAS domain protein 1
Gene Aliases:
HIF-2alpha, HIF2A, HLF, HRF, MOP2, bHLHe73
Chromosome Location:
Chr.17: 86753864 - 86833410 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01236114_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epas1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010137.3 NP_034267.3 9 - 10 1671 62
GenBank mRNA AF045160.1 - 9 - 10 1435 62
AK136911.1 - 9 - 10 1675 62
AK141185.1 - 9 - 10 1406 62
AK158430.1 - 9 - 10 1835 62
BC057870.1 - 9 - 10 1393 5032291 62
D89787.1 - 9 - 10 1633 62
U81983.1 - 9 - 10 1354 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001299.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epas1

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
response to hypoxia
embryonic placenta development
blood vessel remodeling
regulation of heart rate
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to oxidative stress
mitochondrion organization
signal transduction
multicellular organism development
visual perception
hemopoiesis
cell differentiation
erythrocyte differentiation
lung development
norepinephrine metabolic process
surfactant homeostasis
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
positive regulation of transcription from RNA polymerase II promoter
cell maturation
myoblast fate commitment
iron ion homeostasis
cellular response to hypoxia
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein binding
transcription factor binding
histone acetyltransferase binding
sequence-specific DNA binding
protein heterodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700011851.mm.1 NM_010137.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700011851.mm.1 NM_010137.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700011851.mm.1 TC1700001299.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0