Assay Details
| Gene Symbol: Abhd14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC019410.1 | - | 1 - 2 | 49 | 4191819 | 86 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC019410.1 transcripts.
Set Membership: |
|

Abhd14b
Genomic Map