This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235709_m1
See other PPP2R3A GE Assays ›
Gene Symbol:
Ppp2r3a
Gene Name:
protein phosphatase 2, regulatory subunit B'', alpha
Gene Aliases:
3222402P14Rik, A730042E07
Chromosome Location:
Chr.9: 101104989 - 101251832 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235709_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161362.3 NP_001154834.1 2 - 3 2598 109
GenBank mRNA AK076163.1 - 1 - 2 1650 109
BC042841.1 - 1 - 2 1560 109
BC042842.1 - 1 - 2 2033 109
BC151170.1 - 1 - 2 2425 9088059 109
BC151171.1 - 1 - 2 2420 9088060 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

eye photoreceptor cell differentiation
somatic muscle development
positive regulation of protein catabolic process
negative regulation of canonical Wnt signaling pathway
regulation of cell motility involved in somitogenic axis elongation
positive regulation of canonical Wnt signaling pathway
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0