This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235274_m1
See other 4930486I03RIK GE Assays ›
Gene Symbol:
4930486I03Rik
Gene Name:
RIKEN cDNA 4930486I03 gene
Gene Aliases:
-
Chromosome Location:
Chr.1: 20356454 - 20392300 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235274_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930486I03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045734.1 2 - 3 374 125
GenBank mRNA AK015632.1 - 2 - 3 375 125

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream polycystic kidney and hepatic disease 1 Pkhd1 AI118496,
AI182499,
FPC,
Tigm1
Anti-sense Chr. 1 - 20057779 - 20618082 on NCBI Build GRCm38
Downstream microRNA 206 Mir206 Mirn206,
mmu-mir-206
286.7kb Sense Chr. 1 - 20679010 - 20679082 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000124.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100000880.mm.1 NR_045734.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000880.mm.1 NR_045734.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000880.mm.1 TC0100000124.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104920068 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0