This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235255_g1
See other ABCB7 GE Assays ›
Gene Symbol:
Abcb7
Gene Name:
ATP-binding cassette, sub-family B (MDR/TAP), member 7
Gene Aliases:
AA517758, AU019072, Abc7
Chromosome Location:
Chr.X: 104280565 - 104413846 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235255_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcb7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009592.1 NP_033722.1 10 - 11 1388 143
GenBank mRNA AK040069.1 - 10 - 11 1385 143
AK084853.1 - 10 - 11 1379 143
AK151967.1 - 10 - 11 1374 143
AK152535.1 - 10 - 11 1374 143
AK152816.1 - 10 - 11 1374 143
AK152946.1 - 10 - 11 1376 143
BC035534.1 - 7 - 8 1017 143
BC151037.1 - 10 - 11 1388 9087950 143
U43892.1 - 9 - 10 1192 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002797.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
cellular iron ion homeostasis
transmembrane transport
nucleotide binding
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00017644.mm.1 NM_009592.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017644.mm.1 NM_009592.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017644.mm.1 TC0X00002797.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0