This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235211_g1
See other 4930535L15RIK GE Assays ›
Gene Symbol:
4930535L15Rik
Gene Name:
RIKEN cDNA 4930535L15 gene
Gene Aliases:
-
Chromosome Location:
Chr.9: 107998115 - 108002456 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235211_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930535L15Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK019721.1 - 3 - 4 304 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715554.1 - 3 - 4 304 Chr. 9 - 107998116 - 108002404 [-] on Build GRCm38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cadherin-related family member 4 Cdhr4 Cdh29 Anti-sense Chr. 9 - 107992492 - 107999683 on NCBI Build GRCm38
Downstream inositol hexaphosphate kinase 1 Ip6k1 1200016D08Rik,
AU022776,
Ihpk1,
InsP6,
InsP6k1,
Itpk6
3.9kb Anti-sense Chr. 9 - 108002648 - 108048782 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Mouse Genome 430 2.0 Array 1438142_PM_s_at - -
Mouse Genome 430 2.0 Array 1438142_s_at - -
Mouse Expression Set 430 1438142_s_at - -
Mouse Expression Set 430 1438142_PM_s_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0