This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01235086_m1
See other ADHFE1 GE Assays ›
Gene Symbol:
Adhfe1
Gene Name:
alcohol dehydrogenase, iron containing, 1
Gene Aliases:
6330565B14Rik, AI043035, Adh8
Chromosome Location:
Chr.1: 9547969 - 9577968 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01235086_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adhfe1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175236.4 NP_780445.1 9 - 10 923 82
NR_027664.1 8 - 9 882 82
GenBank mRNA AK018241.1 - 9 - 10 894 82
AK038853.1 - 9 - 10 890 82
AK046780.1 - 9 - 10 849 82
AK050178.1 - 8 - 9 847 82
AK084104.1 - 852 82
BC026584.1 - 9 - 10 881 4972323 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000039.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adhfe1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

molecular hydrogen transport
oxidation-reduction process
oxidoreductase activity
metal ion binding
hydroxyacid-oxoacid transhydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100000307.mm.1 NM_175236.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000307.mm.1 NM_175236.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100000307.mm.1 NR_027664.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000307.mm.1 NR_027664.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100000307.mm.1 TC0100000039.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0