This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01234672_m1
See other GPR183 GE Assays ›
Gene Symbol:
Gpr183
Gene Name:
G protein-coupled receptor 183
Gene Aliases:
Ebi2
Chromosome Location:
Chr.14: 121952331 - 121965193 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01234672_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr183
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183031.2 NP_898852.2 1 - 2 142 111
GenBank mRNA AK087951.1 - 1 - 2 55 111
AK138693.1 - 1 - 2 144 111
AK141593.1 - 1 - 2 54 111
AK153216.1 - 1 - 2 55 111
AK156005.1 - 1 - 2 111
AK156289.1 - 1 - 2 61 111
BC052868.1 - 1 - 2 49 30073287 111

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK138693.1, AK153216.1, AK156005.1, BC052868.1, NM_183031.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr183

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

mature B cell differentiation involved in immune response
immune system process
humoral immune response
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of B cell proliferation
positive regulation of ERK1 and ERK2 cascade
signal transducer activity
G-protein coupled receptor activity
oxysterol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400020476.mm.1 NM_183031.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400020476.mm.1 NM_183031.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400020476.mm.1 TC1400002746.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400020477.mm.1 NM_183031.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400020477.mm.1 NM_183031.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400020477.mm.1 TC1400002746.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400020478.mm.1 NM_183031.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400020478.mm.1 NM_183031.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400020478.mm.1 TC1400002746.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0