This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01234360_g1
See other NONO GE Assays ›
Gene Symbol:
Nono
Gene Name:
non-POU-domain-containing, octamer binding protein
Gene Aliases:
AA407051, AV149256, NRB54, P54NRB, nonA
Chromosome Location:
Chr.X: 101429560 - 101448593 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01234360_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nono
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252518.1 NP_001239447.1 3 - 4 337 95
NM_023144.2 NP_075633.2 3 - 4 340 95
GenBank mRNA AK004830.1 - 3 - 4 303 95
AK013444.1 - 3 - 4 254 95
AK028338.1 - 3 - 4 305 95
AK033201.1 - 3 - 4 266 95
AK145179.1 - 3 - 4 313 95
AK161152.1 - 3 - 4 324 95
AK165855.1 - 3 - 4 299 95
AK167297.1 - 3 - 4 271 95
AK169202.1 - 3 - 4 303 95
AK169303.1 - 3 - 4 324 95
AK213551.1 - 1 - 2 163 95
BC005465.1 - 3 - 4 299 3600935 95
BC083074.1 - 1 - 2 177 6390386 95
CJ180326.1 - 3 - 4 340 95
CT010241.1 - 1 - 2 159 95
S64860.1 - 3 - 4 287 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nono

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
cellular response to DNA damage stimulus
circadian rhythm
RNA splicing
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
rhythmic process
negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
nucleotide binding
core promoter binding
nucleic acid binding
DNA binding
RNA binding
protein binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header