This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233663_m1
See other MRPL15 GE Assays ›
Gene Symbol:
Mrpl15
Gene Name:
mitochondrial ribosomal protein L15
Gene Aliases:
HSPC145, MRP-L7, Rpml7
Chromosome Location:
Chr.1: 4773198 - 4785745 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233663_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177658.1 NP_001171129.1 3 - 4 477 93
NM_025300.4 NP_079576.1 3 - 4 477 93
NR_033530.1 2 - 3 322 93
GenBank mRNA AK003367.1 - 3 - 4 461 93
AK010474.1 - 3 - 4 437 93
AK010604.1 - 3 - 4 470 93
AK011775.1 - 2 - 3 297 93
AK017804.1 - 3 - 4 454 93
AK017824.1 - 3 - 4 440 93
AK019175.1 - 3 - 4 450 93
AK019244.1 - 3 - 4 437 93
AK019249.1 - 3 - 4 437 93
AK150880.1 - 3 - 4 478 93
AK160660.1 - 3 - 4 435 93
BC021304.1 - 3 - 4 473 3498041 93
BC027233.1 - 3 - 4 462 3498065 93
BC068230.1 - 3 - 4 460 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001909.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrpl15

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial genome maintenance
translation
structural constituent of ribosome
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0