This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01233628_m1
See other SEMA3E GE Assays ›
Gene Symbol:
Sema3e
Gene Name:
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E
Gene Aliases:
6430702L12, AA408817, Semah, mKIAA0331
Chromosome Location:
Chr.5: 14025276 - 14256689 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01233628_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema3e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011348.2 NP_035478.2 12 - 13 2090 109
GenBank mRNA AF034744.1 - 12 - 13 1825 109
AK129116.1 - 7 - 8 855 109
AK141295.1 - 11 - 12 2812 110
AK142340.1 - 12 - 13 1917 109
AK159214.1 - 12 - 13 1881 109
BC057956.1 - 12 - 13 1988 5357516 109
Z80941.1 - 12 - 13 1456 109
Z93947.1 - 12 - 13 2068 109
Z93948.1 - 12 - 13 2068 109

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema3e

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
patterning of blood vessels
negative regulation of cell-matrix adhesion
sprouting angiogenesis
multicellular organism development
nervous system development
regulation of cell shape
negative regulation of angiogenesis
cell differentiation
synapse organization
semaphorin-plexin signaling pathway
regulation of actin cytoskeleton reorganization
protein binding
semaphorin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0