This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232956_m1
See other CAMKMT GE Assays ›
Gene Symbol:
Camkmt Gnao1
Gene Name:
calmodulin-lysine N-methyltransferase
guanine nucleotide binding protein, alpha O
Gene Aliases:
1700106N22Rik, AI480743, AV099781, CLNMT, CaM KMT
AW050213, Galphao, Gnao, alphaO
Chromosome Location:
Chr.17: 85090592 - 85458589 on Build GRCm38
Chromosome Location:
Chr.8: 93810561 - 93969388 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm01232956_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camkmt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028576.2 NP_082852.1 2 - 3 340 81
GenBank mRNA AK007130.1 - 2 - 3 435 81
AK155246.1 - 2 - 3 341 81
AK172547.1 - 2 - 3 286 81
BC138272.1 - 2 - 3 488 8861291 81
BC138273.1 - 2 - 3 488 8861292 81
Gene Symbol: Gnao1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK147783.1 - 377 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001270.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
methylation
molecular_function
methyltransferase activity
transferase activity
calmodulin-lysine N-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700011738.mm.1 NM_028576.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700011738.mm.1 NM_028576.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700011738.mm.1 TC1700001270.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0