This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232792_g1
See other GPR108 GE Assays ›
Gene Symbol:
Gpr108
Gene Name:
G protein-coupled receptor 108
Gene Aliases:
1810015L19Rik, AA589464, C79132, Lustr2
Chromosome Location:
Chr.17: 57234914 - 57248441 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232792_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr108
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF376726.1 - 10 - 11 1061 69
AK019012.1 - 1 - 2 35 69
AK047364.1 - 10 - 11 1037 69
AK082797.1 - 10 - 11 1087 69
AK159313.1 - 10 - 11 1106 69
AX775895.1 - 10 - 11 1118 69
AX775897.1 - 10 - 11 1061 69
BC016104.1 - 10 - 11 1087 4920840 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK019012.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700022132.mm.1 NM_030084.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700022132.mm.1 TC1700002354.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0