This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232594_g1
See other RRP9 GE Assays ›
Gene Symbol:
Rrp9
Gene Name:
RRP9, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases:
55kDa, D19435, D9Wsu10e, Rnu3ip2, U3-55k
Chromosome Location:
Chr.9: 106477309 - 106485415 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232594_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rrp9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145620.4 NP_663595.1 10 - 11 1010 65
GenBank mRNA AF368232.1 - 10 - 11 993 65
AK076113.1 - 10 - 11 1012 65
AK156199.1 - 9 - 10 1084 65
BC014703.1 - 10 - 11 1010 4237895 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001350.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rrp9

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
biological_process
RNA binding
snoRNA binding
U3 snoRNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900010559.mm.1 NM_145620.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010559.mm.1 NM_145620.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900010559.mm.1 TC0900001350.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0