This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01232055_m1
See other IP6K2 GE Assays ›
Gene Symbol:
Ip6k2
Gene Name:
inositol hexaphosphate kinase 2
Gene Aliases:
1500005N04Rik, AW050208, Ihpk2
Chromosome Location:
Chr.9: 108783764 - 108806333 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01232055_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ip6k2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029634.2 NP_083910.2 3 - 4 694 137
GenBank mRNA AK050593.1 - 3 - 4 550 137
BC039922.1 - 3 - 4 705 2609803 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001422.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ip6k2

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphate ion transport
phosphorylation
negative regulation of cell growth
positive regulation of apoptotic process
phosphatidylinositol phosphorylation
nucleotide binding
inositol hexakisphosphate 5-kinase activity
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
kinase activity
transferase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900011525.mm.1 NM_029634.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011525.mm.1 NM_029634.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011525.mm.1 TC0900001422.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900011514.mm.1 NM_029634.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011514.mm.1 NM_029634.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011514.mm.1 TC0900001422.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900011522.mm.1 NM_029634.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011522.mm.1 NM_029634.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011522.mm.1 TC0900001422.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900011513.mm.1 NM_029634.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011513.mm.1 NM_029634.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011513.mm.1 TC0900001422.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900011524.mm.1 NM_029634.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011524.mm.1 NM_029634.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011524.mm.1 TC0900001422.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900011523.mm.1 NM_029634.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011523.mm.1 NM_029634.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011523.mm.1 TC0900001422.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0