This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01231894_g1
See other PCBP4 GE Assays ›
Gene Symbol:
Pcbp4
Gene Name:
poly(rC) binding protein 4
Gene Aliases:
1200003L19Rik, AlphaCP-4
Chromosome Location:
Chr.9: 106453406 - 106464012 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01231894_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcbp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021567.5 NP_067542.3 4 - 5 378 62
GenBank mRNA AF176328.1 - 4 - 5 264 62
AK004580.1 - 4 - 5 329 62
AK005383.1 - 4 - 5 327 62
AK082005.1 - 4 - 5 328 62
BC010694.1 - 4 - 5 267 2654291 62
BY259572.1 - 4 - 5 365 62
CJ162439.1 - 4 - 5 375 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001349.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcbp4

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of mRNA splicing, via spliceosome
nucleic acid binding
DNA binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900010526.mm.1 NM_021567.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010526.mm.1 NM_021567.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900010526.mm.1 TC0900001349.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900010525.mm.1 NM_021567.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010525.mm.1 NM_021567.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900010525.mm.1 TC0900001349.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0