This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01231859_m1
See other MRAS GE Assays ›
Gene Symbol:
Mras
Gene Name:
muscle and microspikes RAS
Gene Aliases:
2900078C09Rik, AI326250
Chromosome Location:
Chr.9: 99385420 - 99436961 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01231859_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mras
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008624.3 NP_032650.1 1 - 2 406 83
GenBank mRNA AB004879.1 - 1 - 2 81
AK050469.1 - 1 - 2 250 83
CJ162751.1 - 1 - 2 395 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB004879.1, AK050469.1, NM_008624.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mras

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
small GTPase mediated signal transduction
nucleotide binding
GTP binding
GTP-dependent protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900024091.mm.1 NM_008624.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900024091.mm.1 NM_008624.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900024091.mm.1 TC0900002921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900024088.mm.1 NM_008624.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900024088.mm.1 NM_008624.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900024088.mm.1 TC0900002921.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 567402 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0