Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900009573.mm.1 |
NM_001185164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009573.mm.1 |
NM_001185164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009573.mm.1 |
NM_011697.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009573.mm.1 |
NM_011697.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009573.mm.1 |
TC1900001027.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900009575.mm.1 |
NM_001185164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009575.mm.1 |
NM_001185164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009575.mm.1 |
NM_011697.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009575.mm.1 |
NM_011697.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900009575.mm.1 |
TC1900001027.mm.1 |
MTA TC |