Assay Details
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166368.1 | NP_001159840.1 | 5 - 6 | 433 | 105 | |
NM_008020.3 | NP_032046.1 | 5 - 6 | 448 | 105 | ||
GenBank mRNA | AK165394.1 | - | 5 - 6 | 413 | 105 | |
BC031824.1 | - | 5 - 6 | 402 | 4196363 | 105 | |
BC053692.1 | - | 5 - 6 | 438 | 6395841 | 105 | |
CF949699.1 | - | 4 - 5 | 252 | 105 | ||
DV655383.1 | - | 5 - 6 | 433 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937107 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937112 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937110 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937103 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937105 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937111 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937108 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937106 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937109 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | CF949699.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | DV655383.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | BC031824.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | BC053692.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | CF949699.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | DV655383.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | BC031824.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | BC053692.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859627 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | CF949699.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | DV655383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | BC031824.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | BC053692.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | CF949699.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | DV655383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | BC031824.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | BC053692.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | CF949699.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | DV655383.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | BC031824.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | BC053692.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | CF949699.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | DV655383.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | BC031824.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | BC053692.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P291588 | CF949699.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P291588 | DV655383.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P291588 | BC031824.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P291588 | BC053692.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P291588 | NM_001166368.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937090 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937117 | - | - |
Whole Mouse Genome G4122A | A_51_P464234 | CF949699.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | DV655383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | BC031824.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | BC053692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | CF949699.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | DV655383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | BC031824.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | BC053692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464234 | NM_001166368.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P291588 | CF949699.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P291588 | DV655383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P291588 | BC031824.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P291588 | BC053692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P291588 | NM_001166368.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112813 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14227649 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14227647 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14311554 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937108 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937106 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937103 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937109 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937111 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937105 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937112 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937110 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14287635 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14311564 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937093 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14503898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14287637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14227647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14311535 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937086 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14314373 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31937097 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10465555 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465554 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465558 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465557 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465556 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450694_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10465553 | - | - |
Mouse Genome 430 2.0 Array | 1450694_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450694_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14227_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14227_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900009555.mm.1 | NM_001166368.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009555.mm.1 | NM_001166368.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009555.mm.1 | NM_008020.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009555.mm.1 | NM_008020.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009555.mm.1 | TC1900001026.mm.1 | MTA TC |
Mouse Expression Set 430 | 1450694_at | - | - |
Mouse Expression Set 430 | 1450694_PM_at | - | - |
Murine Genome U74v2 Set | 99546_PM_at | - | - |
Murine Genome U74v2 Set | 99546_at | - | - |
Mouse Genome 430A 2.0 Array | 1450694_at | - | - |
Mouse Genome 430A 2.0 Array | 1450694_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450694_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450694_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 919163 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2320025 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002320025 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220875 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1220875 | - | - |
mouse-6 v1.1 (Illumina) | scl014227.3_23-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2320025 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl014227.3_23-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320025 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2962273 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1220875 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220875 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2962273 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014227.3_23-S | - | - |
