This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01231326_g1
See other PPP1CA GE Assays ›
Gene Symbol:
Ppp1ca
Gene Name:
protein phosphatase 1, catalytic subunit, alpha isoform
Gene Aliases:
Ppp1c, dism2
Chromosome Location:
Chr.19: 4192174 - 4195419 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01231326_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1ca
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031868.2 NP_114074.1 3 - 4 453 76
GenBank mRNA AK007932.1 - 3 - 4 454 76
AK028392.1 - 3 - 4 443 76
AK090070.1 - 3 - 4 471 76
AK150344.1 - 3 - 4 471 76
AK151582.1 - 3 - 4 471 76
AK152051.1 - 3 - 4 471 76
AK152585.1 - 3 - 4 471 76
AK152667.1 - 3 - 4 449 76
AK153517.1 - 3 - 4 435 76
AK158681.1 - 3 - 4 409 76
AK159575.1 - 3 - 4 461 76
AK167244.1 - 3 - 4 454 76
AK167538.1 - 3 - 4 466 76
AK167880.1 - 3 - 4 455 76
AK167981.1 - 3 - 4 454 76
BC014828.1 - 3 - 4 459 4239005 76
U25809.1 - 3 - 4 474 76

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1ca

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycogen metabolic process
regulation of glycogen biosynthetic process
regulation of glycogen catabolic process
regulation of translation
protein dephosphorylation
cell cycle
female meiotic division
cellular process
dephosphorylation
lung development
circadian regulation of gene expression
regulation of circadian rhythm
entrainment of circadian clock by photoperiod
rhythmic process
branching morphogenesis of an epithelial tube
cell division
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
protein binding
protein phosphatase 1 binding
hydrolase activity
phosphatase activity
protein complex binding
ribonucleoprotein complex binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000472.mm.1 NM_031868.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000472.mm.1 NM_031868.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000472.mm.1 TC1900000033.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header