This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01231212_g1
See other TCIRG1 GE Assays ›
Gene Symbol:
Tcirg1
Gene Name:
T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3
Gene Aliases:
ATP6N1C, ATP6a3, Atp6i, OC-116, OPTB1, Stv1, TIRC7, Vph1, oc
Chromosome Location:
Chr.19: 3896050 - 3907133 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01231212_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcirg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136091.2 NP_001129563.1 15 - 16 2008 61
NM_001167784.1 NP_001161256.1 15 - 16 2353 61
NM_016921.3 NP_058617.3 15 - 16 2049 61
GenBank mRNA AB022322.1 - 15 - 16 1915 61
AF218253.1 - 15 - 16 1987 61
AF218254.1 - 8 - 9 1670 61
AK004094.1 - 6 - 7 859 61
AK155140.1 - 15 - 16 2356 61
BC006761.1 - 15 - 16 2049 3257437 61
BC085234.1 - 15 - 16 1909 5322429 61

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcirg1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

vacuolar acidification
ATP synthesis coupled proton transport
vacuolar proton-transporting V-type ATPase complex assembly
proton-transporting ATPase activity, rotational mechanism
ATPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0