This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230921_g1
See other MTA2 GE Assays ›
Gene Symbol:
Mta2
Gene Name:
metastasis-associated gene family, member 2
Gene Aliases:
AW550797, Mata1l1, Mta1l1, mmta2
Chromosome Location:
Chr.19: 8941920 - 8952300 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230921_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mta2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011842.3 NP_035972.3 10 - 11 1394 102
GenBank mRNA AF159259.1 - 10 - 11 1269 102
AK077548.1 - 10 - 11 1376 102
AK088158.1 - 10 - 11 1307 102
AK147099.1 - 10 - 11 1373 102
AK149834.1 - 10 - 11 1440 102
AK157717.1 - 10 - 11 1298 102
AK159979.1 - 10 - 11 1370 102
AK182140.1 - 1 - 2 89 102
BC079847.1 - 10 - 11 1394 6419411 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000168.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mta2

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA methylation
DNA packaging
chromatin remodeling
regulation of transcription, DNA-templated
regulation of fibroblast migration
histone deacetylation
ATP-dependent chromatin remodeling
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcription factor activity, transcription factor binding
RNA polymerase II transcription factor binding
RNA polymerase II repressing transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
histone deacetylase activity
protein binding
zinc ion binding
nucleosomal DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002299.mm.1 NM_011842.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002299.mm.1 NM_011842.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900002299.mm.1 TC1900000168.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900002298.mm.1 NM_011842.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002298.mm.1 NM_011842.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900002298.mm.1 TC1900000168.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0