Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500000757.mm.1 |
NM_001243072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000757.mm.1 |
NM_001243072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000757.mm.1 |
NM_001243073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000757.mm.1 |
NM_001243073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000757.mm.1 |
NM_009152.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000757.mm.1 |
NM_009152.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000757.mm.1 |
TC0500000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000758.mm.1 |
NM_001243072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000758.mm.1 |
NM_001243072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000758.mm.1 |
NM_001243073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000758.mm.1 |
NM_001243073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000758.mm.1 |
NM_009152.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000758.mm.1 |
NM_009152.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000758.mm.1 |
TC0500000084.mm.1 |
MTA TC |