This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01230644_g1
See other SEMA3F GE Assays ›
Gene Symbol:
Sema3f
Gene Name:
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F
Gene Aliases:
Sema4, Semak
Chromosome Location:
Chr.9: 107681499 - 107710475 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01230644_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema3f
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011349.3 NP_035479.2 13 - 14 1651 91
GenBank mRNA AF080090.1 - 13 - 14 1557 91
AF080091.1 - 12 - 13 1450 91
AK053632.1 - 13 - 14 1652 91
BC010976.1 - 13 - 14 1572 4219060 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003042.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema3f

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
axon guidance
facial nerve structural organization
trigeminal nerve structural organization
nerve development
branchiomotor neuron axon guidance
ventral trunk neural crest cell migration
negative regulation of axon extension involved in axon guidance
axon extension involved in axon guidance
negative chemotaxis
sympathetic ganglion development
sympathetic neuron projection extension
sympathetic neuron projection guidance
neural crest cell migration involved in autonomic nervous system development
semaphorin-plexin signaling pathway involved in neuron projection guidance
semaphorin-plexin signaling pathway involved in axon guidance
chemorepellent activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900025439.mm.1 NM_011349.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900025439.mm.1 NM_011349.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900025439.mm.1 TC0900003042.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900025440.mm.1 NM_011349.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900025440.mm.1 NM_011349.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900025440.mm.1 TC0900003042.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0