Assay Details
Gene Symbol: Nprl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018879.2 | NP_061367.1 | 10 - 11 | 1242 | 76 | |
GenBank mRNA | AF131207.1 | - | 10 - 11 | 1241 | 76 | |
AK010331.1 | - | 10 - 11 | 1160 | 76 | ||
AK013299.1 | - | 10 - 11 | 1204 | 76 | ||
AK013691.1 | - | 10 - 11 | 1193 | 76 | ||
AK088925.1 | - | 8 - 9 | 1532 | 76 | ||
BC026548.1 | - | 10 - 11 | 1158 | 5028027 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AK088925.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | BC026548.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AF131207.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AK010331.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AK013299.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AK088925.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | BC026548.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AF131207.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AK010331.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59457 | AK013299.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796291 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860677 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444396 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177215 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AK088925.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | BC026548.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AF131207.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AK010331.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AK013299.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AK088925.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | BC026548.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AF131207.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AK010331.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59457 | AK013299.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809422 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809446 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809442 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809424 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177215 | - | - |
Whole Mouse Genome G4122A | A_52_P59457 | AK088925.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | BC026548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | AF131207.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | AK010331.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | AK013299.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | AK088925.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | BC026548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | AF131207.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | AK010331.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59457 | AK013299.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137817 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128012 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177215 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07487478 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07487462 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07683233 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07487459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07487462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809447 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07934884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26809443 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10588644 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588645 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588647 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588649 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588648 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588651 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588643 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588654 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588650 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588652 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588653 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417046_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10588642 | - | - |
Mouse Genome 430 2.0 Array | 1417046_at | - | - |
Mouse Genome 430 2.0 Array | 1417046_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56032_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56032_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900010762.mm.1 | NM_018879.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900010762.mm.1 | NM_018879.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900010762.mm.1 | TC0900001371.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417046_at | - | - |
Mouse Expression Set 430 | 1417046_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417046_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417046_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417046_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417046_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6100113 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006100113 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2454195 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2454195 | - | - |
mouse-6 v1.1 (Illumina) | scl36496.9.1_37-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6100113 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl36496.9.1_37-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6100113 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2454195 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2454195 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36496.9.1_37-S | - | - |
